siRNA Details
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011563.5 | NP_035693.3 | 2 | 188 | ||
GenBank mRNA | AK005225.1 | - | 2 | 216 | ||
AK008433.1 | - | 2 | 193 | |||
AK010653.1 | - | 2 | 163 | |||
AK011963.1 | - | 136 | ||||
AK014765.1 | - | 1 | 146 | |||
AK088280.1 | - | 2 | 207 | |||
BC002034.1 | - | 2 | 196 | 3485635 | ||
BC081454.1 | - | 2 | 107 | 6818396 | ||
BC086783.1 | - | 1 | 74 | 5700069 | ||
U20611.1 | - | 2 | 150 | |||
U51679.1 | - | 2 | 125 | |||
X82067.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
