siRNA Details
Gene Symbol: Dntt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043228.1 | NP_001036693.1 | 3 | 641 | ||
NM_009345.2 | NP_033371.2 | 3 | 641 | |||
GenBank mRNA | AF316014.1 | - | 3 | 494 | ||
AF316015.1 | - | 3 | 494 | |||
AK039804.1 | - | 3 | 532 | |||
AK087978.1 | - | 3 | 642 | |||
AK088709.1 | - | 3 | 642 | |||
AK134009.1 | - | 3 | 535 | |||
X04123.1 | - | 3 | 528 | |||
X68670.2 | - | 3 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
