siRNA Details
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 2 | 341 | ||
NM_153484.3 | NP_705617.2 | 2 | 248 | |||
GenBank mRNA | AF194420.1 | - | 2 | 275 | ||
AK042146.1 | - | 2 | 502 | |||
AK045575.1 | - | 2 | 197 | |||
AK053229.1 | - | 2 | 186 | |||
AK141606.1 | - | 2 | 169 | |||
AK145125.1 | - | 2 | 310 | |||
AK147965.1 | - | 2 | 274 | |||
AK154472.1 | - | 2 | 286 | |||
AK177827.1 | - | 2 | 52 | |||
AK181094.1 | - | 2 | 52 | |||
AK185529.1 | - | 2 | 52 | |||
AK185648.1 | - | 1 | 52 | |||
AK214482.1 | - | 2 | 52 | |||
AY540631.1 | - | 2 | 327 | |||
AY540632.1 | - | 2 | 142 | |||
BC017689.1 | - | 2 | 104 | 4242534 | ||
BC036982.1 | - | 2 | 305 | 4976241 | ||
BY296040.1 | - | 2 | 248 |
