siRNA Details
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148926.2 | NP_683728.1 | 3 | 357 | ||
GenBank mRNA | AK082915.1 | - | 2 | 353 | ||
AK148316.1 | - | 320 | ||||
AK160675.1 | - | 3 | 214 | |||
AK181546.1 | - | 2 | 147 | |||
AK183489.1 | - | 2 | 147 | |||
AK183610.1 | - | 2 | 147 | |||
AK193183.1 | - | 1 | 147 | |||
BC027161.1 | - | 1 | 89 | |||
BC034396.1 | - | 3 | 277 | 3707476 | ||
BC083124.1 | - | 3 | 357 | 6334368 | ||
BC100559.1 | - | 3 | 208 | 3419074 | ||
X80437.1 | - | 3 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
