siRNA Details
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 6, 7 | 854 | ||
GenBank mRNA | AB053464.1 | - | 6, 7 | 799 | ||
AF316616.2 | - | 6, 7 | 711 | |||
AK154050.1 | - | 6, 7 | 830 | |||
AK160587.1 | - | 6, 7 | 795 | |||
AY338234.1 | - | 6, 7 | 784 | |||
BC010716.1 | - | 6, 7 | 767 | 2655861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
