siRNA Details
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019678.3 | NP_062652.1 | 4 | 651 | ||
GenBank mRNA | AK008549.1 | - | 4 | 427 | ||
AK158539.1 | - | 4 | 517 | |||
AK167475.1 | - | 4 | 515 | |||
AK186338.1 | - | 3 | 389 | |||
AK186588.1 | - | 3 | 298 | |||
AK210251.1 | - | 3 | 300 | |||
BC024638.1 | - | 4 | 429 | 4015300 | ||
U94662.1 | - | 4 | 553 |

More Information
Other Species Targeted

Gene Ontology Categories:
