siRNA Details
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009364.3 | NP_033390.1 | 3 | 348 | ||
GenBank mRNA | AK132268.1 | - | 3 | 370 | ||
BC021639.1 | - | 3 | 383 | 4023742 | ||
D50586.1 | - | 3 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
