siRNA Details
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031199.3 | NP_112476.1 | 6 | 1692 | ||
GenBank mRNA | AK162680.1 | - | 6 | 1457 | ||
U65016.1 | - | 6 | 1526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
