siRNA Details
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031199.3 | NP_112476.1 | 3 | 428 | ||
GenBank mRNA | AK010785.1 | - | 3 | 456 | ||
AK162680.1 | - | 3 | 193 | |||
AK166321.1 | - | 3 | 361 | |||
BC132185.1 | - | 3 | 226 | 40130462 | ||
BC132211.1 | - | 3 | 226 | 40130488 | ||
M92420.1 | - | 4 | 178 | |||
U65016.1 | - | 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
