siRNA Details
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177751.1 | NP_001171222.1 | 2 | 260 | ||
NM_009366.3 | NP_033392.1 | 2 | 352 | |||
NM_207652.2 | NP_997535.2 | 2 | 3209 | |||
GenBank mRNA | AF201285.1 | - | 2 | 2920 | ||
AK135277.1 | - | 2 | 349 | |||
AK135282.1 | - | 2 | 350 | |||
AK135514.1 | - | 2 | 349 | |||
AK136211.1 | - | 2 | 157 | |||
AK145204.1 | - | 2 | 351 | |||
AK147039.1 | - | 2 | 353 | |||
AK148732.1 | - | 2 | 350 | |||
AK173322.1 | - | 4 | 3621 | |||
AK179000.1 | - | 2 | 299 | |||
BC026703.1 | - | 2 | 297 | |||
BC058660.1 | - | 2 | 3209 | 6409347 | ||
BY720238.1 | - | 2 | 151 | |||
X62940.1 | - | 2 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
