siRNA Details
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009371.3 | NP_033397.3 | 4 | 833 | ||
NM_029575.3 | NP_083851.3 | 3 | 758 | |||
GenBank mRNA | AF406755.1 | - | 3 | 432 | ||
AK043619.1 | - | 4 | 917 | |||
AK046561.1 | - | 4 | 759 | |||
AK053793.1 | - | 4 | 834 | |||
AK148167.1 | - | 3 | 758 | |||
AK159327.1 | - | 4 | 813 | |||
BC052629.1 | - | 735 | ||||
S69114.1 | - | 3 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
