siRNA Details
Gene Symbol: Tgtp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145164.1 | NP_001138636.1 | 2 | 782 | ||
GenBank mRNA | AK088858.1 | - | 2 | 776 | ||
AK172580.1 | - | 2 | 775 | |||
L38444.1 | - | 2 | 613 |
Gene Symbol: Tgtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011579.3 | NP_035709.3 | 2 | 775 | ||
GenBank mRNA | AK089836.1 | - | 2 | 777 | ||
AK163978.1 | - | 2 | 776 | |||
AK172473.1 | - | 2 | 775 | |||
BC034256.1 | - | 2 | 740 | 3662733 | ||
BC085259.1 | - | 746 | 5389279 | |||
BC093522.1 | - | 2 | 757 | 3988138 | ||
CT010183.1 | - | 1 | 514 | |||
U15636.1 | - | 2 | 619 |
