siRNA Details
Gene Symbol: Timeless | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136082.2 | NP_001129554.1 | 20, 21 | 2694 | ||
NM_001164080.1 | NP_001157552.1 | 20, 21 | 2694 | |||
NM_001164081.1 | NP_001157553.1 | 20, 21 | 2694 | |||
NM_011589.2 | NP_035719.1 | 20, 21 | 2637 | |||
GenBank mRNA | AB015598.1 | - | 19, 20 | 2514 | ||
AB019001.2 | - | 20, 21 | 2665 | |||
AF071506.1 | - | 20, 21 | 2665 | |||
AF098161.1 | - | 20, 21 | 2648 | |||
AF126480.1 | - | 4, 5 | 541 | |||
AK153838.1 | - | 20, 21 | 2669 | |||
AK162012.1 | - | 19, 20 | 2818 | |||
BC026526.1 | - | 6, 7 | 763 | |||
BC058641.1 | - | 20, 21 | 2626 | 6837105 | ||
BC082770.1 | - | 20, 21 | 2623 | 6314901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
