siRNA Details
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044384.1 | NP_001037849.1 | 2 | 212 | ||
NM_001294280.2 | NP_001281209.1 | 2 | 212 | |||
NM_011593.2 | NP_035723.2 | 2 | 209 | |||
GenBank mRNA | AA184230.1 | - | 1 | 58 | ||
BC008107.1 | - | 2 | 114 | 3158222 | ||
BC034260.1 | - | 2 | 114 | 3158222 | ||
BC051260.1 | - | 2 | 94 | 30056565 | ||
CJ055052.1 | - | 2 | 124 | |||
CJ063120.1 | - | 2 | 210 | |||
M17243.1 | - | 2 | 75 | |||
X04684.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
