siRNA Details
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198985.1 | NP_001185914.1 | 21 | 3376 | ||
NM_011597.4 | NP_035727.2 | 21 | 3366 | |||
GenBank mRNA | AF113005.1 | - | 21 | 3224 | ||
AK028494.1 | - | 21 | 3294 | |||
AK184185.1 | - | 1 | 112 | |||
AK209802.1 | - | 1 | 149 | |||
BC034677.1 | - | 21 | 3284 | 3155078 | ||
BC039924.1 | - | 21 | 3295 | 3155079 | ||
BC042695.1 | - | 21 | 3268 | |||
CF736959.1 | - | 2 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
