siRNA Details
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198985.1 | NP_001185914.1 | 13 | 1956 | ||
NM_011597.4 | NP_035727.2 | 13 | 1946 | |||
GenBank mRNA | AF113005.1 | - | 13 | 1804 | ||
AK028494.1 | - | 13 | 1875 | |||
BC034677.1 | - | 13 | 1864 | 3155078 | ||
BC039924.1 | - | 13 | 1875 | 3155079 | ||
BC042695.1 | - | 13 | 1848 | |||
CN527767.1 | - | 5 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
