siRNA Details
Gene Symbol: Dtymk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023136.2 | NP_075625.1 | 2 | 582 | ||
GenBank mRNA | AA014471.1 | - | 1 | 348 | ||
AK011955.1 | - | 2 | 525 | |||
AK019284.1 | - | 2 | 517 | |||
AK019288.1 | - | 2 | 556 | |||
BC069902.1 | - | 2 | 553 | 5026527 | ||
CF983443.1 | - | 2 | 570 | |||
S82852.1 | - | 2 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
