siRNA Details
Gene Symbol: Tnfsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011613.3 | NP_035743.2 | 1, 2 | 381 | ||
GenBank mRNA | AB008426.1 | - | 1, 2 | 227 | ||
AB032771.1 | - | 2, 3 | 140 | |||
AB036798.1 | - | 1, 2 | 227 | |||
AF013170.1 | - | 1, 2 | 368 | |||
AF019048.1 | - | 1, 2 | 363 | |||
AF053713.1 | - | 1, 2 | 396 | |||
AK159498.1 | - | 1, 2 | 382 | |||
AK159997.1 | - | 1, 2 | 382 | |||
BC125603.1 | - | 1, 2 | 284 | 40130118 | ||
BC131970.1 | - | 1, 2 | 284 | 40130247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
