siRNA Details
Gene Symbol: Cd70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011617.2 | NP_035747.1 | 1 | 303 | ||
GenBank mRNA | BC125407.1 | - | 1 | 262 | 40129922 | |
BC125409.1 | - | 1 | 262 | 40129924 | ||
U78091.1 | - | 1 | 185 | |||
Y13636.1 | - | 1 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
