siRNA Details
Gene Symbol: Tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009405.2 | NP_033431.1 | 4 | 156 | ||
GenBank mRNA | BC028515.1 | - | 5 | 194 | 1448494 | |
J04992.1 | - | 4 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
