siRNA Details
Gene Symbol: Tnni3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009406.3 | NP_033432.1 | 4 | 258 | ||
GenBank mRNA | AK169223.1 | - | 4 | 259 | ||
BC061171.1 | - | 4 | 258 | 30300830 | ||
BC100590.1 | - | 4 | 189 | 6787916 | ||
U09181.1 | - | 2 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
