siRNA Details
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277903.1 | NP_001264832.1 | 10 | 525 | ||
NM_001277904.1 | NP_001264833.1 | 9 | 489 | |||
NM_011618.2 | NP_035748.1 | 10 | 522 | |||
GenBank mRNA | AF020946.1 | - | 10 | 509 | ||
AJ131711.1 | - | 9 | 470 | |||
AK003317.1 | - | 2 | 104 | |||
AK160226.1 | - | 10 | 523 | |||
BC138871.1 | - | 10 | 502 | 8861893 | ||
BC141142.1 | - | 10 | 505 | 9055754 | ||
BC145449.1 | - | 9 | 469 | 9054007 | ||
BC145450.1 | - | 9 | 469 | 9054008 | ||
BC145451.1 | - | 10 | 505 | 9054009 | ||
U92883.1 | - | 10 | 506 | |||
U92884.1 | - | 9 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
