siRNA Details
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011623.2 | NP_035753.2 | 18 | 2197 | ||
GenBank mRNA | AK033321.1 | - | 6 | 579 | ||
AK169723.1 | - | 3 | 209 | |||
D12513.1 | - | 18 | 2123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
