siRNA Details
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025261.1 | NP_001020432.1 | 4 | 488 | ||
NM_001025262.1 | NP_001020433.1 | 4 | 488 | |||
NM_001025263.1 | NP_001020434.1 | 4 | 391 | |||
NM_001025264.1 | NP_001020435.1 | 4 | 391 | |||
NM_009412.2 | NP_033438.1 | 4 | 391 | |||
GenBank mRNA | AK005456.1 | - | 4 | 372 | ||
AK032111.1 | - | 4 | 354 | |||
AK150935.1 | - | 4 | 354 | |||
AK151471.1 | - | 4 | 352 | |||
AK151744.1 | - | 4 | 352 | |||
AK152543.1 | - | 4 | 352 | |||
AK153266.1 | - | 4 | 352 | |||
AK153292.1 | - | 4 | 362 | |||
AK161753.1 | - | 4 | 353 | |||
AK168379.1 | - | 4 | 347 | |||
AK168817.1 | - | 4 | 355 | |||
AK169751.1 | - | 4 | 384 | |||
AK170725.1 | - | 4 | 337 | |||
AK171050.1 | - | 4 | 332 | |||
AY048852.1 | - | 4 | 480 | |||
BC002036.1 | - | 4 | 371 | 3593963 | ||
BE634665.1 | - | 4 | 373 | |||
BG404433.1 | - | 4 | 483 | |||
CA480795.1 | - | 4 | 525 | |||
U44426.1 | - | 4 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
