siRNA Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009413.1 | NP_033439.1 | 3 | 431 | ||
GenBank mRNA | AF004428.1 | - | 3 | 431 | ||
AK131855.1 | - | 3 | 328 | |||
BC141068.1 | - | 3 | 318 | 9055680 | ||
BC145321.1 | - | 3 | 318 | 9053876 | ||
BC145322.1 | - | 3 | 318 | |||
BC171924.1 | - | 3 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
