siRNA Details
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155434.1 | - | 7 | 968 | ||
BC060705.1 | - | 8 | 1189 | 6409657 | ||
D84655.1 | - | 8 | 1042 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155434.1 | - | 7 | 968 | ||
BC060705.1 | - | 8 | 1189 | 6409657 | ||
D84655.1 | - | 8 | 1042 |