siRNA Details
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133977.2 | NP_598738.1 | 16 | 2034 | ||
GenBank mRNA | AF440692.1 | - | 15 | 1964 | ||
AK085754.1 | - | 16 | 2019 | |||
AK142599.1 | - | 2024 | ||||
AK146549.1 | - | 16 | 2019 | |||
AK149559.1 | - | 16 | 2019 | |||
AK149595.1 | - | 16 | 2019 | |||
AK150782.1 | - | 16 | 2067 | |||
AK168368.1 | - | 16 | 2021 | |||
AK168405.1 | - | 16 | 2021 | |||
AK168419.1 | - | 16 | 2019 | |||
AK168938.1 | - | 16 | 2019 | |||
AK169187.1 | - | 16 | 2018 | |||
BC008559.1 | - | 14 | 1756 | |||
BC012313.1 | - | 16 | 2034 | 3981648 | ||
BC018573.1 | - | 11 | 1304 | |||
BC020295.1 | - | 16 | 2003 | |||
BC022986.1 | - | 16 | 2012 | 5123741 | ||
BC058216.1 | - | 16 | 2000 | |||
BC058218.1 | - | 16 | 2013 | |||
BC092046.1 | - | 16 | 1995 | 3594705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
