siRNA Details
Gene Symbol: Trhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013696.2 | NP_038724.1 | 2 | 911 | ||
GenBank mRNA | AK134591.1 | - | 2 | 912 | ||
BC109005.1 | - | 2 | 796 | 40048176 | ||
BC109006.1 | - | 2 | 796 | 40048178 | ||
BC128269.1 | - | 2 | 796 | 40048433 | ||
M59811.1 | - | 2 | 820 | |||
M94384.1 | - | 2 | 820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
