siRNA Details
Gene Symbol: Xntrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011644.3 | NP_035774.2 | 11 | 1814 | ||
GenBank mRNA | AK040538.1 | - | 11 | 2155 |
Gene Symbol: Trpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109897.2 | NP_001103367.1 | 2 | 650 | ||
GenBank mRNA | AF230803.1 | - | 2 | 650 |

More Information
Other Species Targeted

Gene Ontology Categories:
