siRNA Details
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 9 | 2528 | ||
GenBank mRNA | AF029983.1 | - | 9 | 2183 | ||
AF060107.1 | - | 9 | 2527 | |||
AJ006204.1 | - | 8 | 2125 | |||
AK047867.1 | - | 7 | 1549 | |||
BC112972.1 | - | 9 | 2346 | 40049027 |

More Information
Other Species Targeted

Gene Ontology Categories:
