siRNA Details
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 6 | 1950 | ||
GenBank mRNA | AF029983.1 | - | 6 | 1605 | ||
AF060107.1 | - | 6 | 1949 | |||
AJ006204.1 | - | 5 | 1547 | |||
AK038531.1 | - | 6 | 1774 | |||
AK047867.1 | - | 4 | 971 | |||
BC112972.1 | - | 6 | 1768 | 40049027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
