siRNA Details
Gene Symbol: Trpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282086.1 | NP_001269015.1 | 2 | 1145 | ||
NM_001282087.1 | NP_001269016.1 | 2 | 1145 | |||
NM_013838.2 | NP_038866.2 | 2 | 1145 | |||
GenBank mRNA | AF057748.1 | - | 2 | 714 | ||
AK133864.1 | - | 2 | 1045 | |||
AK137447.1 | - | 1 | 375 | |||
BC067391.1 | - | 2 | 1091 | |||
BC068310.1 | - | 2 | 1091 | |||
BC141131.1 | - | 2 | 1016 | 9055743 | ||
U49069.1 | - | 2 | 1145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
