siRNA Details
Gene Symbol: Trpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282086.1 | NP_001269015.1 | 6 | 1878 | ||
NM_013838.2 | NP_038866.2 | 7 | 2112 | |||
GenBank mRNA | AF057748.1 | - | 7 | 1681 | ||
AK133864.1 | - | 6 | 1778 | |||
AK137447.1 | - | 6 | 1340 | |||
BC141131.1 | - | 7 | 1983 | 9055743 | ||
U49069.1 | - | 7 | 2112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
