siRNA Details
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 6 | 690 | ||
NM_001286713.1 | NP_001273642.1 | 6 | 690 | |||
NM_001286714.1 | NP_001273643.1 | 6 | 685 | |||
NM_001286716.1 | NP_001273645.1 | 6 | 685 | |||
NM_001286718.1 | NP_001273647.1 | 6 | 690 | |||
NM_001286720.1 | NP_001273649.1 | 6 | 690 | |||
NM_011647.3 | NP_035777.2 | 6 | 690 | |||
GenBank mRNA | AF132986.1 | - | 2 | 218 | ||
AK143799.1 | - | 6 | 691 | |||
AK147016.1 | - | 6 | 666 | |||
AK147302.1 | - | 6 | 630 | |||
AK147487.1 | - | 6 | 659 | |||
AK147874.1 | - | 6 | 655 | |||
AK147908.1 | - | 6 | 691 | |||
AK148062.1 | - | 6 | 682 | |||
AK148072.1 | - | 6 | 587 | |||
AK170589.1 | - | 6 | 681 | |||
AK171007.1 | - | 6 | 659 | |||
AK190229.1 | - | 3 | 280 | |||
AK193509.1 | - | 3 | 281 | |||
AK214850.1 | - | 3 | 258 | |||
AK217069.1 | - | 3 | 280 | |||
BC052449.1 | - | 6 | 666 | 6409037 | ||
BC060701.1 | - | 6 | 666 | 6409037 | ||
U37775.1 | - | 5 | 602 | |||
U39818.1 | - | 6 | 661 |
