siRNA Details
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113404.1 | NP_001106875.1 | 5 | 546 | ||
NM_011648.5 | NP_035778.3 | 5 | 546 | |||
GenBank mRNA | AK014519.1 | - | 5 | 545 | ||
AK021020.1 | - | 5 | 540 | |||
AK029084.1 | - | 5 | 512 | |||
AK041086.1 | - | 5 | 523 | |||
BB645263.1 | - | 5 | 546 | |||
BC028754.1 | - | 5 | 501 | |||
BC086691.1 | - | 5 | 475 | 30682686 | ||
BC092523.1 | - | 5 | 488 | 30650002 | ||
U02601.1 | - | 5 | 528 | |||
U02602.1 | - | 5 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
