siRNA Details
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011648.5 | NP_035778.3 | 10 | 1063 | ||
GenBank mRNA | AK014519.1 | - | 10 | 1062 | ||
AK029084.1 | - | 10 | 1029 | |||
BC086691.1 | - | 10 | 992 | 30682686 | ||
BC092523.1 | - | 10 | 1005 | 30650002 | ||
U02601.1 | - | 10 | 1045 | |||
U02602.1 | - | 10 | 1045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
