siRNA Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011650.3 | NP_035780.1 | 2 | 148 | ||
GenBank mRNA | AF234179.1 | - | 2 | 148 | ||
AK010737.1 | - | 2 | 146 | |||
AK033003.1 | - | 1 | 388 | |||
AK150529.1 | - | 2 | 197 | |||
AK151448.1 | - | 2 | 199 | |||
AK165598.1 | - | 2 | 188 | |||
AK181409.1 | - | 2 | 114 | |||
AK183078.1 | - | 2 | 114 | |||
AK184665.1 | - | 2 | 118 | |||
AK198107.1 | - | 2 | 114 | |||
AK199692.1 | - | 2 | 114 | |||
AK202525.1 | - | 2 | 114 | |||
BC004615.1 | - | 2 | 194 | 3585366 | ||
X81464.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
