siRNA Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011650.3 | NP_035780.1 | 3 | 255 | ||
GenBank mRNA | AF234179.1 | - | 3 | 255 | ||
AK010737.1 | - | 3 | 253 | |||
AK033003.1 | - | 1 | 1427 | |||
AK150529.1 | - | 3 | 304 | |||
AK151448.1 | - | 3 | 306 | |||
AK165598.1 | - | 3 | 295 | |||
AK181409.1 | - | 3 | 221 | |||
AK183078.1 | - | 3 | 221 | |||
AK184665.1 | - | 3 | 225 | |||
AK189586.1 | - | 3 | 221 | |||
AK198107.1 | - | 3 | 220 | |||
AK199692.1 | - | 3 | 221 | |||
AK202525.1 | - | 3 | 221 | |||
BC004615.1 | - | 3 | 301 | 3585366 | ||
X81464.1 | - | 3 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
