siRNA Details
Gene Symbol: Tspyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009433.3 | NP_033459.1 | 1 | 924 | ||
GenBank mRNA | AF042180.1 | - | 1 | 888 | ||
AK053812.1 | - | 1 | 875 | |||
AK159432.1 | - | 1 | 905 | |||
AK166438.1 | - | 1 | 905 | |||
AK166805.1 | - | 1 | 853 | |||
AK166893.1 | - | 1 | 853 | |||
BC011213.1 | - | 1 | 871 | 4196841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
