siRNA Details
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077591.1 | NP_001071059.1 | 3 | 504 | ||
NM_011651.2 | NP_035781.2 | 3 | 504 | |||
GenBank mRNA | AF025310.2 | - | 3 | 484 | ||
BC118618.1 | - | 3 | 483 | 40107271 | ||
BC121827.1 | - | 3 | 483 | 40107270 | ||
CN831975.1 | - | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
