siRNA Details
Gene Symbol: Tst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009437.4 | NP_033463.1 | 1 | 197 | ||
GenBank mRNA | AK003953.1 | - | 1 | 204 | ||
BC005644.1 | - | 1 | 221 | 3257034 | ||
CT010210.1 | - | 1 | 171 | |||
U35741.1 | - | 1 | 185 |

More Information
Other Species Targeted

Gene Ontology Categories:
