siRNA Details
Gene Symbol: Tsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009440.2 | NP_033466.1 | 5 | 378 | ||
GenBank mRNA | AK018925.1 | - | 1 | 19 | ||
BC048493.1 | - | 5 | 378 | 6705048 | ||
X99796.1 | - | 5 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
