siRNA Details
Gene Symbol: Tgoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009443.3 | NP_033469.1 | 2 | 1107 | ||
GenBank mRNA | AK041302.1 | - | 2 | 857 | ||
AK076586.1 | - | 2 | 1108 | |||
AK133559.1 | - | 2 | 855 | |||
AK161242.1 | - | 2 | 840 | |||
AK162574.1 | - | 2 | 857 | |||
BC009143.1 | - | 2 | 842 | 3157708 | ||
BC141218.1 | - | 2 | 852 | 9055830 | ||
BC145542.1 | - | 2 | 852 | 9054105 | ||
CT010269.1 | - | 2 | 829 | |||
D50031.1 | - | 2 | 855 |
