siRNA Details
Gene Symbol: Ttr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013697.5 | NP_038725.1 | 3 | 457 | ||
GenBank mRNA | AK014454.1 | - | 3 | 294 | ||
AK018701.1 | - | 3 | 295 | |||
AK050155.1 | - | 3 | 294 | |||
AK075588.1 | - | 3 | 294 | |||
BC024702.1 | - | 3 | 427 | 4192268 | ||
BC032069.1 | - | 3 | 284 | |||
BC086926.1 | - | 3 | 294 | 6749152 | ||
CK627038.1 | - | 3 | 279 | |||
D89076.1 | - | 3 | 294 | |||
X03351.1 | - | 3 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
