siRNA Details
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011653.2 | NP_035783.1 | 4 | 1534 | ||
GenBank mRNA | AK077529.1 | - | 4 | 1545 | ||
AK145404.1 | - | 4 | 1583 | |||
AK146983.1 | - | 4 | 1583 | |||
AK161783.1 | - | 2 | 1130 | |||
AK164335.1 | - | 4 | 1585 | |||
AK167687.1 | - | 4 | 1583 | |||
AK168762.1 | - | 4 | 1583 | |||
AK169610.1 | - | 4 | 1584 | |||
BC056169.1 | - | 4 | 1545 | 5715646 | ||
BC083343.1 | - | 4 | 1558 | 5687414 | ||
BC083344.1 | - | 4 | 1553 | 5693701 | ||
BC083345.1 | - | 4 | 1568 | 6415067 | ||
BC085256.1 | - | 4 | 1567 | 6306481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
