siRNA Details
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011653.2 | NP_035783.1 | 4 | 720 | ||
GenBank mRNA | AK077529.1 | - | 4 | 732 | ||
AK145404.1 | - | 4 | 769 | |||
AK146983.1 | - | 4 | 769 | |||
AK164335.1 | - | 4 | 771 | |||
AK167687.1 | - | 4 | 769 | |||
AK168762.1 | - | 4 | 769 | |||
AK169610.1 | - | 4 | 770 | |||
BC056169.1 | - | 4 | 731 | 5715646 | ||
BC083343.1 | - | 4 | 744 | 5687414 | ||
BC083344.1 | - | 4 | 739 | 5693701 | ||
BC083345.1 | - | 4 | 754 | 6415067 | ||
BC085256.1 | - | 4 | 753 | 6306481 | ||
M13445.1 | - | 4 | 720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
