siRNA Details
Gene Symbol: Tulp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011657.2 | NP_035787.1 | 5 | 516 | ||
GenBank mRNA | AB016963.1 | - | 5 | 459 | ||
AF045582.1 | - | 5 | 469 | |||
BC060068.1 | - | 5 | 446 | 6419643 |

More Information
Other Species Targeted

Gene Ontology Categories:
