siRNA Details
Gene Symbol: ITGAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002209.2 | NP_002200.2 | 5 | 537 | ||
XM_005255313.1 | XP_005255370.1 | 5 | 457 | |||
XM_011545849.1 | XP_011544151.1 | 5 | 470 | |||
GenBank mRNA | AK298674.1 | - | 5 | 454 | ||
AK314246.1 | - | 5 | 491 | |||
DC324373.1 | - | 5 | 537 | |||
Y00796.1 | - | 5 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
