siRNA Details
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 1 | 366 | ||
GenBank mRNA | AK014619.1 | - | 1 | 406 | ||
AY526904.1 | - | 1 | 407 | |||
BC079678.1 | - | 1 | 316 | 30539846 | ||
D00131.1 | - | 1 | 369 | |||
D00440.1 | - | 1 | 366 | |||
M24560.1 | - | 1 | 366 | |||
M26729.1 | - | 1 | 305 | |||
X12782.1 | - | 1 | 407 |
