siRNA Details
Gene Symbol: ITGAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114380.1 | NP_001107852.1 | 3, 4 | 436 | ||
NM_002209.2 | NP_002200.2 | 3, 4 | 436 | |||
XM_005255313.1 | XP_005255370.1 | 3, 4 | 356 | |||
XM_006721044.1 | XP_006721107.1 | 3, 4 | 387 | |||
XM_011545849.1 | XP_011544151.1 | 3, 4 | 369 | |||
GenBank mRNA | AK298674.1 | - | 3, 4 | 353 | ||
AK314246.1 | - | 3, 4 | 390 | |||
BC008777.2 | - | 3, 4 | 369 | 3142951 | ||
DC324373.1 | - | 3, 4 | 436 | |||
Y00796.1 | - | 3, 4 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
